missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Ovine Prolactin Antagonist Protein

A cDNA sequence encoding the Prolactin Antagonist was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP14010-EC-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15915409

  • 137.00€ / 10 microgramos
Fecha estimada de envío: 29-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Especificaciones

Especificaciones

Prolactin Antagonist Protein
Research Use Only
Ovine Prolactin Antagonist is devoid of agonistic activity and capable of inhibiting biological activity of Ovine Prolactin or other lactogenic hormones as evidenced by proliferation assay of Nb2 or other cells.
Ovine
His
Greater than 99.0% as determined by SDS-PAGE.
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
Ovine Prolactin was lyophilized from a concentrated (1 mg/ml) solution with 0.02%-0.03% NaHCO3.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales