missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Ovine Leptin tA Protein

A cDNA sequence encoding the Leptin tA was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10767-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15915108

  • 103.00€ / 5 microgramos


Fecha de envío estimada
del proveedor 05-04-2019
Añadir a la cesta

Especificación

Especificación

Leptin tA Protein
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
The protein was lyophilized from a concentrated (0.65 mg/ml) solution with 0.003mM NaHCO3.
Greater than 95.0% as determined by: (a) Gel filtration analysis. (b) Analysis by SDS-PAGE.
Research Use Only
Pegylated triple antagonist is capable of inhibiting leptin-induced proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. Its in vitro activity is 6-8 fold lower than the non-pegylated antagonist but in vivo it has profound weight gain effect (as compared to the non-pegylated antagonist like in mouse leptin antagonists), resulting mainly from increased food intake.
Ovine
Untagged
Videos