missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Uricase Protein
A cDNA sequence encoding the Uricase was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10908-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Uricase Protein | |
Research Use Only | |
The specific activity was found to be 10U/mg. One Unit oxidizes one micromole of uric acid per minute at 25C, at pH 8.5. | |
Other | |
Untagged | |
Each 1.5 mg Uricase contains 5 mg sucrose, 25 mg glycine, 0.1 mg Tween-80, 13.6 mg Na2HPO4*12H20 and 0.33 mg NaH2PO4*2H20. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
msavkaaryg kdnvrvykvh kdektgvqtv yemtvcvlle geietsytka dnsvivatds ikntiyitak qnpvtppelf gsilgthfie kynhihaahv nivchrwtrm didgkphphs firdseekrn vqvdvvegkg idiksslsgl tvlkstnsqf wgflrdeytt lketwdrils tdvdatwqwk nfsglqevrs hvpkfdatwa tarevtlktf aednsasvqa tmykmaeqil arqqlietve yslpnkhyfe idlswhkglq ntgknaevfa pqsdpnglik ctvgrsslks kl | |
Greater than 90.0% as determined by SDS-PAGE. |