missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Treponema p17 16.4kDa Protein
A cDNA sequence encoding the Treponema p17 16.4kDa was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13809-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Treponema p17 16.4kDa Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Lyophilized from 1 mg/ml in 20mM sodium carbonate pH 10. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Greater than 95.0% as determined by SDS-PAGE. |