missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Thymosin ?4 Protein
A cDNA sequence encoding the Thymosin ?4 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13746-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Thymosin ?4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
SDKPDMAEIE KFDKSKLKKT ETQEKNPLPS KETIEQEKQA GES | |
Greater than 95.0% as determined by SDS-PAGE and HPLC analyses. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Lyophilized from a 0.2um filtered concentrated solution in 1xPBS, pH 7.4. |