missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other TBEV NE Protein

A cDNA sequence encoding the TBEV NE was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13701-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15924429

  • 1141.00€ / 1 miligramo


Fecha de envío estimada
del proveedor 03-04-2019
Añadir a la cesta

Especificación

Especificación

TBEV NE Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Encephalitis protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
20mM MES pH 6.5, 8M urea, 200 mM NaCl and 0.05% Tween-20.
Videos