missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other T. Vaginalis P65 Protein

A cDNA sequence encoding the T. Vaginalis P65 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13679-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15924359

  • 183.00€ / 100 microgramos


Fecha estimada de envío
23-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

T. Vaginalis P65 Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Protein is >90% pure as determined by 12% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
T. Vaginalis p65protein solution in PBS and 2M Urea.
Videos