missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other T. pallidum TmpA partial Protein

A cDNA sequence encoding the T. pallidum TmpA partial was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13678-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914359

  • 691.00€ / 500 microgramos


Fecha de envío estimada
del proveedor 05-04-2019
Añadir a la cesta

Especificación

Especificación

T. pallidum TmpA partial Protein
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
10mM Tris-HCl pH 8.0, 20mM DDT, 1mM EDTA and 8M urea.
Videos