missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other T. pallidum p47 Protein
A cDNA sequence encoding the T. pallidum p47 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13675-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
T. pallidum p47 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
8M urea, 10mM Tris-HCl pH 8.0, 1mM EDTA and 1mM DTT. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Treponema Pallidum protein is >95% pure as determined by SDS-PAGE. |