missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other SPA 33.4kDa Protein
A cDNA sequence encoding the SPA 33.4kDa was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13567-100mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SPA 33.4kDa Protein | |
Research Use Only | |
spa | |
Other | |
Untagged | |
SPA protein was lyophilized with no additives. |
100 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
NAAQHDEAQQ NAFYQVLNMP NLNADQRNGF IQSLKDDPSQ SANVLGEAQK LNDSQAPKAD AQQNNFNKDQ QSAFYEILNM PNLNEAQRNG FIQSLKDDPS QSTNVLGEAK KLNESQAPKA DNNFNKEQQN AFYEILNMPN LNEEQRNGFI QSLKDDPSQS ANLLSEAKKL NESQAPKADN KFNKEQQNAF YEILHLPNLN EEQRNGFIQS LKDDPSQSAN LLAEAKKLND AQAPKADNKF NKEQQNAFYE ILHLPNLTEE QRNGFIQSLK DDPSVSKEIL AEAKKLNDAQ APKEED | |
Greater than 97.0% as determined by: (a) Analysis by HPLC. (b) Analysis by SDS-PAGE. |