missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Shiga Like Toxin 2 Protein

A cDNA sequence encoding the Shiga Like Toxin 2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13496-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923759

  • 1372.00€ / 1 miligramo


Fecha estimada de envío
18-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

Shiga Like Toxin 2 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
Shiga Like Toxin 2 protein solution contains PBS, 50mM arginine and 0.05% sodium azide.
Videos