missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other scIC Protein

A cDNA sequence encoding the scIC was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13432-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923549

  • 103.00€ / 2 microgramos


Fecha de envío estimada
del proveedor 10-04-2019
Añadir a la cesta

Especificación

Especificación

scIC Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Greater than 85.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
Each mg/ml of protein contains 50% glycerol.
Videos