missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other S. Pyogenes tkt Protein

A cDNA sequence encoding the S. Pyogenes tkt was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13381-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923389

Cantidad Precio
1 1065.00€ / 100 microgramos
4 992.47€ / 100 microgramos
Fecha estimada de envío
09-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

S. Pyogenes tkt Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Greater than 80.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
S. Pyogenes tkt is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
Videos
Documentos