missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other RNASEH1 Protein
A cDNA sequence encoding the RNASEH1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13325-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
RNASEH1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMLKQVEI FTDGSCLGNP GPGGYGAILR YRGREKTFSA GYTRTTNNRM ELMAAIVALE ALKEHCEVIL STDSQYVRQG ITQWIHNWKK RGWKTADKKP VKNVDLWQRL DAALGQHQIK WEWVKGHAGH PENERCDELA RAAAMNPTLE DTGYQVEV | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
RNASEH1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 2mM DTT. |