missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other protein 1

A cDNA sequence encoding the Protein A was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13160-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15922689

  • 103.00€ / 10 microgramos


Fecha estimada de envío
21-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

protein 1
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MAQHDEAQQN AFYQVLNMPN LNADQRNGFI QSLKDDPSQS ANVLGEAQKL NDSQAPKADA QQNNFNKDQQ SAFYEILNMP NLNEAQRNGFIQSLKDDPSQ STNVLGEAKK LNESQAPKAD NNFNKEQQNA FYEILNMPNL NEEQRNGFIQ SLKDDPSQSA NLLSEAKKLN ESQAPKADNKFNKEQQNAFY EILHLPNLNE EQRNGFIQSL KDDPSQSANL LAEAKKLNDA QAPKADNKFN KEQQNAFYEI LHLPNLTEEQ RNGFIQSLKDDPSVSKEILA EAKKLNDAQA PKEEDNNKPG KEDNNKPGKE DNNKPGKEDG NKPGKEDNKK PGKEDNKKPG KEDNKKPGKE DGNKPGKEDNKKPGKEDGNG VHVVKPGDTV NDIAKANGTT ADKIAADNKL ADKNMIKPGQ ELVVDKKQPA NHADANKAQA LPET
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
spa
Other
Untagged
The Protein-A protein solution contains 20mM Tris-HCl, pH 8 and 10% glycerol.
Videos
Documentos