missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Protease Protein

A cDNA sequence encoding the Protease was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13159-50IU

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15912689

  • 103.00€ / 50 unidades


Fecha estimada de envío
20-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

Protease Protein
Research Use Only
Recombinant Protein
E. coli
50 IU
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Videos
Documentos