missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other NusA Protein
A cDNA sequence encoding the NusA was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12911-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NusA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MNKEILAVVE AVSNEKALPR EKIFEALESA LATATKKKYE QEIDVRVQID RKSGDFDTFRRWLVVDEVTQ PTKEITLEAA RYEDESLNLG DYVEDQIESV TFDRITTQTA KQVIVQKVREAERAMVVDQF REHEGEIITG VVKKVNRDNI SLDLGNNAEA VILREDMLPR ENFRPGDRVR GVLYSVRPEA RGAQLFVTRS KPEMLIELFR IEVPEIGEEV IEIKAAARDP GSRAKIAVKT NDKRIDPVGA CVGMRGARVQ AVSTELGGER IDIVLWDDNP AQFVINAMAP ADVASIVVDE DKHTMDIAVE AGNLAQAIGR NGQNVRLASQ LSGWELNVMT DDLQAKHQA EAHAAIDTFT KYLDIDEDFA TVLVEEGFST LEELAYVPMK ELLEIEGLDE PTVEALRERA KNALATIAQA QEESLGDNKP ADDLLNLEGV DRDLAFKLAA RGVCTLEDLA EQGIDDLADI EGLTDEKAGA LIMAARNICW FGDEA | |
Greater than 95.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
NusA protein solution contains 1x PBS pH 7.4. |