missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other MMLV RT Protein

A cDNA sequence encoding the MMLV RT was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12705-50000U

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15921289

Cantidad Precio
1 710.00€ / 50000 unidades
5 661.51€ / 50000 unidades
Fecha estimada de envío
07-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

MMLV RT Protein
Research Use Only
Recombinant Protein
E. coli
50mM Tris-HCl, 0.1M NaCl, 0.1% Triton X-100, 2mM DTT, 0.1mM EDTA and 50% glycerol.
50000 IU
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Videos
Documentos