missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other M. Pneumoniae P116 Protein

A cDNA sequence encoding the M. Pneumoniae P116 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12613-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15920999

  • 1372.00€ / 1 miligramo


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

M. Pneumoniae P116 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
GST
Greater than 95% as determined by 12% PAGE (Coomassie staining).
Research Use Only
Recombinant Protein
E. coli
Mycoplasma Pneumoniae P116 is formulated in 1xPBS pH 7.4 and 0.5mM EDTA.
Videos