missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other M. Pneumoniae P116 Protein
A cDNA sequence encoding the M. Pneumoniae P116 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12613-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
M. Pneumoniae P116 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Mycoplasma Pneumoniae P116 is formulated in 1xPBS pH 7.4 and 0.5mM EDTA. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
GST | |
Greater than 95% as determined by 12% PAGE (Coomassie staining). |