missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other GrpE Protein
A cDNA sequence encoding the GrpE was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12053-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
GrpE Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSSKEQKTPE GQAPEEIIMD QHEEIEAVEP EASAEQVDPR DEKIANLEAQ LAEAQTRERD GILRVKAEME NLRRRTELDI EKAHKFALEK FINELLPVID SLDRALEVAD KANPDMSAMV EGIELTLKSM LDVVRKFGVE VIAETNVPLD PNVHQAIAMV ESDDVAPGNV LGIMQKGYTL NGRTIRAAMV TVAKAKA | |
Greater than 90.0% as determined by analysis by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The HSP24 protein solution contains 20mM Tris-HCl pH 8 and 0.1M NaCl. |