missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other GrpE Protein

A cDNA sequence encoding the GrpE was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12053-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919218

  • 103.00€ / 5 microgramos


Fecha de envío estimada
del proveedor 05-04-2019
Añadir a la cesta

Especificación

Especificación

GrpE Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
The HSP24 protein solution contains 20mM Tris-HCl pH 8 and 0.1M NaCl.
Greater than 90.0% as determined by analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MSSKEQKTPE GQAPEEIIMD QHEEIEAVEP EASAEQVDPR DEKIANLEAQ LAEAQTRERD GILRVKAEME NLRRRTELDI EKAHKFALEK FINELLPVID SLDRALEVAD KANPDMSAMV EGIELTLKSM LDVVRKFGVE VIAETNVPLD PNVHQAIAMV ESDDVAPGNV LGIMQKGYTL NGRTIRAAMV TVAKAKA
Videos