missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Gliadin Gamma 18.6kDa Wheat Protein
A cDNA sequence encoding the Gliadin Gamma 18.6kDa Wheat was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11984-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Gliadin Gamma 18.6kDa Wheat Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Gliadin is supplied in 20mM HEPES buffer pH 8.0, 200mM NaCl, and 20% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Greater than 94% as determined by SDS-PAGE. |