missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other GCV Protein

A cDNA sequence encoding the GCV was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11945-100mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15908888

  • 114.00€ / 100 miligramos


Fecha estimada de envío
20-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

GCV Protein
100 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Greater than 99.0%.
Research Use Only
Recombinant Protein
E. coli
Videos
Documentos