missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Filamin Protein

A cDNA sequence encoding the Filamin was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11883-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918688

  • 160.00€ / 50 microgramos


Fecha de envío estimada
del proveedor 08-04-2019
Añadir a la cesta

Especificación

Especificación

Filamin Protein
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Native Protein
The protein was lyophilized from a 1 mg/ml solution containing 20mM Tris / acetate buffer pH 7.6, 0.1mM EDTA, 2mM DTT, 9M urea and 20mM NaCl.
Videos