missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other EGFP Protein
A cDNA sequence encoding the EGFP was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11759-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
EGFP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MVSKGEELFT GVVPILVELD GDVNGHKFSV SGEGEGDATY GKLTLKFICT TGKLPVPWPT LVTTLTYGVQ CFSRYPDHMK QHDFFKSAMP EGYVQERTIF FKDDGNYKTR AEVKFEGDTL VNRIELKGID FKEDGNILGH KLEYNYNSHN VYIMADKQKN GIKVNFKIRH NIEDGSVQLA DHYQQNTPIG DGPVLLPDNH YLSTQSALSK DPNEKRDHMV LLEFVTAAGI TLGMDELYK | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The EGFP was lyophilized from a 0.2microm filtered concentrated solution in PBS pH 7.4. |