missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other DsbC Protein

A cDNA sequence encoding the DsbC was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11704-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918098

  • 1603.00€ / 1 miligramo


Fecha de envío estimada
del proveedor 04-04-2019
Añadir a la cesta

Especificación

Especificación

947363
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
dsbC
Other
Untagged
1 mg/ml solution containing 20mM phosphate buffer pH 7.5 and 20mM EDTA.
DsbC Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
DDAAIQQTLA KMGIKSSDIQ PAPVAGMKTV LTNSGVLYIT DDGKHIIQGP MYDVSGTAPVNVTNKMLLKQ LNALEKEMIV YKAPQEKHVI VFTDITCGY CHKLHEQMAD YNALGITVRYLAFPRQGLDS DAEKEMKAIW CAKDKNKAFD DVMAGKSVAP ASCDVDIADH YVLGVQLGVSGTPAVVLSNG TLVPGYQPPK EMKEFLDEHQ KMTSGK
Videos