missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other BSA Protein

A cDNA sequence encoding the BSA was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11206-10gr

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15936478

  • 171.00€ / 10 g


Fecha estimada de envío
18-12-2019
Añadir a la cesta

Especificaciones

Especificaciones

280717
Greater than 97.0%.
Research Use Only
ALB
Other
Untagged
BSA Protein
10 g
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Native Protein
MKWVTFISLL LLFSSAYSRG VFRRDTHKSE IAHRFKDLGE EHFKGLVLIA FSQYLQQCPF DEHVKLVNEL TEFAKTCVAD ESHAGCEKSL HTLFGDELCK VASLRETYGD MADCCEKQEP ERNECFLSHK DDSPDLPKLK PDPNTLCDEF KADEKKFWGK YLYEIARRHP YFYAPELLYY ANKYNGVFQE CCQAEDKGAC LLPKIETMRE KVLASSARQR LRCASIQKFG ERALKAWSVA RLSQKFPKAE FVEVTKLVTD LTKVHKECCH GDLLECADDR ADLAKYICDN QDTISSKLKE CCDKPLLEKS HCIAEVEKDA IPENLPPLTA DFAEDKDVCK NYQEAKDAFL GSFLYEYSRR HPEYAVSVLL RLAKEYEATL EECCAKDDPH ACYSTVFDKL KHLVDEPQNL IKQNCDQFEK LGEYGFQNAL IVRYTRKVPQ VSTPTLVEVS RSLGKVGTRC CTKPESERMP CTEDYLSLIL NRLCVLHEKT PVSEKVTKCC TESLVNRRPC FSALTPDETY VPKAFDEKLF TFHADICTLP DTEKQIKKQT ALVELLKHKP KATEEQLKTV MENFVAFVDK CCAADDKEAC FAVEGPKLVV STQTALA
Videos
Documentos