missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Borrelia Spielmanii OspC Protein

A cDNA sequence encoding the Borrelia Spielmanii OspC was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11196-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15936448

  • 161.00€ / 20 microgramos


Fecha estimada de envío
30-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

Borrelia Spielmanii OspC Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Greater than 80.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
Borrelia Spielmanii OspC is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
Videos
Documentos