missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Borrelia p41 Protein

A cDNA sequence encoding the Borrelia p41 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11192-HIS-INSECT-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916438

  • 160.00€ / 20 microgramos


Fecha estimada de envío
17-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

Borrelia p41 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Protein is >90% pure as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
25mM glycine, pH 9.6 and 50% glycerol.
Videos