missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Borrelia Afzelii p100 Protein

A cDNA sequence encoding the Borrelia Afzelii p100 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11180-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916398

  • 4118.00€ / 1 miligramo


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

Borrelia Afzelii p100 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Greater than 80.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
Borrelia Afzelii p100 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
Videos