missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Aprotinin Protein

A cDNA sequence encoding the Aprotinin was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11058-250mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916018

  • 310.00€ / 250 miligramos


Fecha de envío estimada
del proveedor 10-04-2019
Añadir a la cesta

Especificación

Especificación

Aprotinin Protein
Research Use Only
Recombinant Protein
Native Protein
MKMSRLCLSV ALLVLLGTLA ASTPGCDTSN QAKAQRPDFC LEPPYTGPCK ARIIRYFYNA KAGLCQTFVY GGCRAKRNNF KSAEDCMRTC GGAIGPWENL
250 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
The protein (1 mg/ml) was lyophilized with no additives.
Videos