missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Antide Protein
A cDNA sequence encoding the Antide was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11028-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Antide Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Ac-D-2-Nal-D-Phe(4-CL)-3-D-Pal-Ser-Lys(nicotinoyl)-D-Lys(nicotinoyl)-Leu-Lys(Isopropyl)-Pro-D-Ala-Nh2 | |
Greater than 98.0% as determined by RP-HPLC. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The Antide peptide was lyophilized with no additives. |