missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Angiostatin K1-4 Protein

A cDNA sequence encoding the Angiostatin K1-4 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10521-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914128

  • 160.00€ / 10 microgramos


Fecha estimada de envío
17-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

Angiostatin K1-4 Protein
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Native Protein
Lyophilized from a (1 mg/ml) solution in containing 20mM Hepes buffer pH 8.2 and 20mM NaCl.
Greater than 98.0% as determined by SDS-PAGE.
Research Use Only
Human Angiostatin Kringles 1-4 significantly inhibits basic-FGF induced endothelial cell proliferation and migration at concentration ranging from 300nM-1.0 uM.
Other
Untagged
Videos