missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse VPS29 Protein

A cDNA sequence encoding the VPS29 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13944-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15935199

  • 6130.00€ / 1 miligramo


Fecha estimada de envío
02-07-2020
Añadir a la cesta

Especificaciones

Especificaciones

VPS29 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
The VPS29 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM Nacl, 1mM DTT and 10% glycerol.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMMLVLV LGDLHIPHRC NSLPAKFKKL LVPGKIQHIL CTGNLCTKES YDYLKTLAGD VHIVRGDFDE NLNYPEQKVV TVGQFKIGLI HGHQVIPWGD MASLALLQRQ FDVDILISGH THKFEAFEHE NKFYINPGSA TGAYNALETN IIPSFVLMDI QASTVVTYVY QLIGDDVKVE RIEYKKS
Videos
Documentos