missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse VEGF Protein
A cDNA sequence encoding the VEGF was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10910-INSECT-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
VEGF Protein | |
Research Use Only | |
The ED50 range, determined by the dose-dependent proliferation of human umbilical vein endothelial cells (HUVEC) (measured by 3H-thymidine uptake) is 1-2 ng/ml, corresponding to a specific activity of 1x106 Units/mg. | |
Mouse | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with no additives. |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Baculovirus | |
MNFLLSWVHW TLALLLYLHH AKWSQAAPTT EGEQKSHEVI KFMDVYQRSY CRPIETLVDI FQEYPDEIEY IFKPSCVPLM RCAGCCNDEA LECVPTSESN ITMQIMRIKP HQSQHIGEMS FLQHSRCECR PKKDRTKPEK KSVRGKGKGQ KRKRKKSRFK SWSVHCEPCS ERRKHLFVQD PQTCKCSCKN TDSRCKARQL ELNERTCRCD KPRR | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |