missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Tyrosine-protein kinase JAK3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Tyrosine-protein kinase JAK3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7690-ye-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001177759.1 | |
Tyrosine-protein kinase JAK3 Protein | |
Research Use Only | |
Jak3 | |
Recombinant Protein | |
Yeast | |
LKYISLLGKG NFGSVELCRY DPLGDNTGPL VAVKQLQHSG PDQQRDFQRE IQILKALHSD FIVKYRGVSY GPGRQSLRLV MEYLPSGCLR DFLQRHRARL HTDRLLLFAW QICKGMEYLG ARRCVHRDLA ARNILVESEA HVKIADFGLA KLLPLGKDYY VVREPGQSPI FWYAPESLSD NIFSRQSDVW SFGVVLYELF TYCDKSCSPS AEFLRMMGPE REGPPLCRLL ELLAEGRRLP PPPTCPTEVQ ELMQLCWAPS PHDRPAFGTL SPQLDALWRG RPG | |
Greater than 90% as determined by SDS-PAGE. |
16453 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |