missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TNF-alpha Protein
A DNA sequence encoding the Mus musculus (Mouse) TNF-alpha, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6811-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001265530.1 | |
TNF-alpha Protein | |
Research Use Only | |
TNF | |
Recombinant Protein | |
E. coli | |
GPQRDEKFPN GLPLISSMAQ TLTLRSSSQN SSDKPVAHVV ANHQVEEQLE WLSQRANALL ANGMDLKDNQ LVVPADGLYL VYSQVLFKGQ GCPDYVLLTH TVSRFAISYQ EKVNLLSAVK SPCPKDTPEG AELKPWYEPI YLGGVFQLEK GDQLSAEVNL PKYLDFAESG QVYFGVIAL | |
Greater than 90% as determined by SDS-PAGE. |
21926 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |