missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Thrombopoietin / THPO Protein

A DNA sequence encoding the Mouse Thrombopoietin / THPO, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10468-ec-250ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923968

  • 1660.00€ / 250 microgramos


Fecha estimada de envío
17-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001166976.1
Thrombopoietin / THPO Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
SPVAPACDPR LLNKLLRDSH LLHSRLSQCP DVDPLSIPVL LPAVDFSLGE WKTQTEQSKA QDILGAVSLL LEGVMAARGQ LEPSCLSSLL GQLSGQVRLL LGALQGLLGT QLPLQGRTTA HKDPNALFLS LQQLLRGKVR FLLLVEGPTL CVRRTLPTTA VPSSTSQLLT LNKF
21832
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
THPO
The ED50 as determined by a cell proliferation assay using human MO7e cells is less than 1.0 ng/ml, corresponding to a specific activity of > 1.0 M 106 IU/mg.
Mouse
Untagged
0.2 um filtered PBS, pH 7.4, with 5% Trehalose, lyophilized
Videos