missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Survivin / BIRC5 / API4 Protein
A DNA sequence encoding the Mus musculus (Mouse) Survivin / BIRC5 / API4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5720-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001012273.1 | |
Survivin / BIRC5 / API4 Protein | |
Research Use Only | |
BIRC5 | |
Recombinant Protein | |
E. coli | |
MGAPALPQIW QLYLKNYRIA TFKNWPFLED CACTPERMAE AGFIHCPTEN EPDLAQCFFC FKELEGWEPD DNPIEEHRKH SPGCAFLTVK KQMEELTVSE FLKLDRQRAK NKIAKETNNK QKEFEETAKT TRQSIEQLAA | |
Greater than 90% as determined by SDS-PAGE. |
11799 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |