missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Stromal cell-derived factor 2-like protein 1

A DNA sequence encoding the Mus musculus (Mouse) Stromal cell-derived factor 2-like protein 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP9916-ye-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16048991

  • 1705.00€ / 500 microgramos


Fecha estimada de envío
02-07-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_071719.1
Stromal cell-derived factor 2-like protein 1
500 μg
Not determined.
Full Length of Mature Protein
Mouse
His
Tris-based buffer, 50% glycerol
64136
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Sdf2l1
Recombinant Protein
Yeast
SKASAGLVTC GSVLKLLNTH HKVRLHSHDI KYGSGSGQQS VTGVEESDDA NSYWRIRGGS EGGCPRGLPV RCGQAVRLTH VLTGKNLHTH HFPSPLSNNQ EVSAFGEDGE GDDLDLWTVR CSGQHWEREA SVRFQHVGTS VFLSVTGEQY GNPIRGQHEV HGMPSANAHN TWKAMEGIFI KPGADLSTGH DEL
Videos
Documentos