missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Stromal cell-derived factor 2-like protein 1
A DNA sequence encoding the Mus musculus (Mouse) Stromal cell-derived factor 2-like protein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9916-ye-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_071719.1 | |
Stromal cell-derived factor 2-like protein 1 | |
Research Use Only | |
Sdf2l1 | |
Recombinant Protein | |
Yeast | |
SKASAGLVTC GSVLKLLNTH HKVRLHSHDI KYGSGSGQQS VTGVEESDDA NSYWRIRGGS EGGCPRGLPV RCGQAVRLTH VLTGKNLHTH HFPSPLSNNQ EVSAFGEDGE GDDLDLWTVR CSGQHWEREA SVRFQHVGTS VFLSVTGEQY GNPIRGQHEV HGMPSANAHN TWKAMEGIFI KPGADLSTGH DEL | |
Greater than 90% as determined by SDS-PAGE. |
64136 | |
10 μg | |
Not determined. | |
Full Length of Mature Protein | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |