missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse STIP1 Protein

A cDNA sequence encoding the STIP1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13616-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914159

  • 160.00€ / 20 microgramos


Fecha estimada de envío
18-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

STIP1 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
The STIP1 solution (1 mg/ml) contains 20mM Tris-HCl(pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MEQVNELKEK GNKALSAGNI DDALQCYSEA IKLDPQNHVL YSNRSAAYAK KGDYQKAYED GCKTVDLKPD WGKGYSRKAA ALEFLNRFEE AKRTYEEGLK HEANNLQLKE GLQNMEARLA ERKFMNPFNL PNLYQKLEND PRTRSLLSDP TYRELIEQLQ NKPSDLGTKL QDPRVMTTLS VLLGVDLGSM DEEEEAATPP PPPPPKKEPK PEPMEEDLPE NKKQALKEKE LGNDAYKKKD FDKALKHYDR AKELDPTNMT YITNQAAVHF EKGDYNKCRE LCEKAIEVGR ENREDYRQIA KAYARIGNSY FKEEKYKDAI HFYNKSLAEH RTPDVLKKCQ QAEKILKEQE RLAYINPDLA LEEKNKGNEC FQKGDYPQAM KHYTEAIKRN PRDAKLYSNR AACYTKLLEF QLALKDCEEC IQLEPTFIKG YTRKAAALEA MKDYTKAMDV YQKALDLDSS CKEAADGYQR CMMAQYNRHD SPEDVKRRAM ADPEVQQIMS DPAMRLILEQ MQKDPQALSE HLKNPVIAQK IQKLMDVGLI AIR
Videos