missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse sRANKL Protein

A cDNA sequence encoding the sRANKL was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10869-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15915418

  • 160.00€ / 10 microgramos


Fecha de envío estimada
del proveedor 11-04-2019
Añadir a la cesta

Especificación

Especificación

sRANKL Protein
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
Lys158-Asp316, with an N-terminal Met, Accession # Q3TWY5
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
The ED50, as measured by its ability to induce osteoclast differentiation of RAW 264.7 mouse monocyte/macrophage cells, is less than 2ng/ml.
Mouse
Untagged
sRANKL protein was lyophilized from a 0.2microm filtered solution in NaH2PO4, NaCl and EDTA.
Videos