missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Serine protease inhibitor Kazal-type 1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Serine protease inhibitor Kazal-type 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7376-ye-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16058550

Cantidad Precio
1 245.00€ / 10 microgramos
6 228.51€ / 10 microgramos
Fecha estimada de envío
16-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_033284.1
Serine protease inhibitor Kazal-type 1 Protein
10 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
20730
Greater than 80% as determined by SDS-PAGE.
Research Use Only
Spink3
Recombinant Protein
Yeast
AKVTGKEASC HDAVAGCPRI YDPVCGTDGI TYANECVLCF ENRKRIEPVL IRKGGPC
Videos
Documentos