missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Serine protease 29 Protein
A DNA sequence encoding the Mus musculus (Mouse) Serine protease 29, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10142-ma-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_444490.2 | |
Serine protease 29 Protein | |
Research Use Only | |
Prss29 | |
Recombinant Protein | |
Mammal | |
GTPAPGPEDV LMGIVGGHSA PQGKWPWQVS LRIYRYYWAF WVHNCGGSII HPQWVLTAAH CIRERDADPS VFRIRVGEAY LYGGKELLSV SRVIIHPDFV HAGLGSDVAL LQLAVSVQSF PNVKPVKLPS ESLEVTKKDV CWVTGWGAVS THRSLPPPYR LQQVQVKIID NSLCEEMYHN ATRHRNRGQK LILKDMLCAG NQGQDSCYGD SGGPLVCNVT GSWTLVGVVS WGYGCALRDF PGVYARVQSF LPWITQQMQR FS | |
Greater than 90% as determined by SDS-PAGE. |
114662 | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |