missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SDF2 Protein
A DNA sequence encoding the Mus musculus (Mouse) SDF2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6658-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_033169.3 | |
SDF2 Protein | |
Research Use Only | |
SDF2 | |
Recombinant Protein | |
E. coli | |
SNMAVVTCGS VVKLLNTRHN VRLHSHDVRY GSGSGQQSVT GVTSVDDSNS YWRIRGKTAT VCERGTPIKC GQPIRLTHIN TGRNLHSHHF TSPLSGNQEV SAFGEEGEGD YLDDWTVLCN GPYWVRDGEV RFKHSSTDVL LSVTGEQYGR PISGQKEVHG MAQPSQNNYW KAMEGIFMKP SELLRAEVHH AEL | |
Greater than 80% as determined by SDS-PAGE. |
20316 | |
10 μg | |
Not determined. | |
Full Length of Mature Protein | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |