missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse SAMHD1 Protein

A DNA sequence encoding the Mus musculus (Mouse) SAMHD1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP9152-ye-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16057721

  • 103.00€ / 10 microgramos


Fecha estimada de envío
18-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001132992.1
SAMHD1 Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
56045
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SAMHD1
Recombinant Protein
Yeast
IDDMEAFTKL TDNIFLEVLH STDPQLSEAQ SILRNIECRN LYKYLGETQP KREKIRKEEY ERLPQEVAKA KPEKAPDVEL KAEDFIVDVI NVDYGMEDKN PIDRVHFYCK SNSKQAVRIN KEQVSQLLPE KFAEQLIRVY CKKKDGKSLD AAGKHFVQWC ALRDFTKPQD GDIIAPLITP LKWNNKTSSC LQEVSKVKTC LKF
Videos
Documentos