missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Resistin Protein

A DNA sequence encoding the Mus musculus (Mouse) Resistin, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6600-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16054460

  • 253.00€ / 50 microgramos


Fecha estimada de envío
21-10-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001191888.1
Resistin Protein
50 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
57264
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RES
Recombinant Protein
E. coli
SSMPLCPIDE AIDKKIKQDF NSLFPNAIKN IGLNCWTVSS RGKLASCPEG TAVLSCSCGS ACGSWDIREE KVCHCQCARI DWTAARCCKL QVAS
Videos
Documentos