missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Properdin Protein

A DNA sequence encoding the Mus musculus (Mouse) Properdin, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5826-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15908049

  • 253.00€ / 50 microgramos


Fecha estimada de envío
16-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_032849.2
Properdin Protein
50 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
18636
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Cfp
Recombinant Protein
E. coli
SDPVLCFTQY EESSGRCKGL LGRDIRVEDC CLNAAYAFQE HDGGLCQACR SPQWSAWSLW GPCSVTCSEG SQLRHRRCVG RGGQCSENVA PGTLEWQLQA CEDQPCCPEM GGWSEWGPWG PCSVTCSKGT QIRQRVCDNP APKCGGHCPG EAQQSQACDT QKTCPTHGAW ASWGPWSPCS GSCLGGAQEP KETRSRSCSA PAPSHQPPGK PCSGPAYEHK ACSGLPPCPV AGGWGPWSPL SPCSVTCGLG QTLEQRTCDH PAPRHGGPFC AGDATRNQMC NKAVPCPVNG EWEAWGKWSD CSRLRMSINC EGTPGQQSRS RSCGGRKFNG KPCAGKLQDI RHCYNIHNCI MKGSWSQWST WSLCTPPCSP NATRVRQRLC TPLLPKYPPT VSMVEGQGEK NVTFWGTPRP LCEALQGQKL VVEEKRSCLH VPVCKDPEEK KP
Videos
Documentos